Merck
  • Electroacupuncture suppresses the pain and pain-related anxiety of chronic inflammation in rats by increasing the expression of the NPS/NPSR system in the ACC.
Electroacupuncture suppresses the pain and pain-related anxiety of chronic inflammation in rats by increasing the expression of the NPS/NPSR system in the ACC.
Brain research (2020-02-12)
Junying Du, Junfan Fang, Zitong Xu, Xuaner Xiang, Sisi Wang, Haiju Sun, Xiaomei Shao, Yongliang Jiang, Boyi Liu, Jianqiao Fang
ABSTRACT
The neuropeptide S/Neuropeptide S receptor (NPS/NPSR) system is involved in the regulation of anxiety in rodents. Chronic inflammation can induce anxiety. Our lab has observed that electroacupuncture (EA) has a beneficial effect on chronic inflammatory pain and pain-related anxiety; however, the mechanism should be further clarified. In the present study, we used an inflammatory pain model to investigate the role of the NPS/NPSR system in the anterior cingulate cortex (ACC) in the analgesic and antianxiety effects of EA. In an inflammatory pain model, the paw withdrawal thresholds (PWTs) were decreased, pain-related anxiety-like behaviors were induced, and the ipsilateral protein expression of NPS and NPSR was decreased in the ACC. EA stimulation increased the PWTs, reduced pain-related anxiety-like behavior, and enhanced the ipsilateral protein expression of NPS and NPSR in the ACC. NPS microinjection increased the PWTs and decreased pain-related anxiety-like behaviors. Furthermore, an NPSR inhibitor combined with EA reversed the effect of EA on the PWTs and pain-related anxiety-like behaviors. Our results suggest that EA suppresses pain and pain-related anxiety-like behavior of chronic inflammation in rats by increasing the expression of the NPS/NPSR system in the ACC.
Sigma-Aldrich
Neuropeptide S from rat, ≥98% (HPLC), powder
Millipore
Immobilon®-P PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.