Merck
Sphingosine in apoptosis signaling.
Biochimica et biophysica acta (2003-01-18)
Olivier Cuvillier
ABSTRACT
The sphingolipid metabolites ceramide, sphingosine, and sphingosine 1-phosphate contribute to controlling cell proliferation and apoptosis. Ceramide and its catabolite sphingosine act as negative regulators of cell proliferation and promote apoptosis. Conversely, sphingosine 1-phosphate, formed by phosphorylation of sphingosine by a sphingosine kinase, has been involved in stimulating cell growth and inhibiting apoptosis. As the phosphorylation of sphingosine diminishes apoptosis, while dephosphorylation of sphingosine 1-phosphate potentiates it, the role of sphingosine as a messenger of apoptosis is of importance. Herein, the effects of sphingosine on diverse signaling pathways implicated in the apoptotic process are reviewed.
Product Description
Avanti
Sphingosine (d22:1), Avanti Research - A Croda Brand 860663P, powder
Avanti
Sphingosine (d14:1), Avanti Research - A Croda Brand 860670P, powder
Avanti
Sphingosine (d16:1), Avanti Research - A Croda Brand 860669P, powder
Avanti
sphingosine-d7, Avanti Research - A Croda Brand 860657P, powder