Merck
alpha-Gal epitopes in animal tissue glycoproteins and glycolipids.
Sub-cellular biochemistry (1999-07-07)
L Rydberg, J Holgersson, B E Samuelsson, M E Breimer
ABSTRACT
alpha-Gal terminated saccharides are present on the cell surface both as glycolipids and glycoproteins in all mammals except Old World monkeys and humans. The structural diversity among identified saccharides terminated by this epitope in animal tissues is steadily increasing. The majority of these saccharides have the alpha-Gal linked to lactosamine but other core saccharides exist. The alpha-Gal terminated saccharides are recognized by the immune system as a specific antigen and antibodies directed to the alpha-Gal, which do not cross-react with the classic blood group B trisaccharide, are found in man and Old World monkeys. Similar to other complex carbohydrate cell surface antigens, the alpha-Gal epitope is heterogeneously distributed in different organs and in different cells within an organ. It is present on the vascular endothelium and it is the primary target for human naturally occurring antibodies following pig to primate/man xenotransplantation leading to hyperacute rejection of the graft. Important for the future will be to further structurally characterize this antigen system, its cellular/subcellular distribution, and to identify possible of additional glycosyltransferases, related to the already described alpha 1,3galactosyltransferase that may explain the structural diversity. Such information will be of importance in the studies of, for example, the pathogenesis of autoimmune diseases and for the production of genetically modified pigs to prevent xenograft rejection.