Merck
  • Modulating inflammatory macrophages with an apoptotic body-inspired nanoparticle.
Modulating inflammatory macrophages with an apoptotic body-inspired nanoparticle.
Acta biomaterialia (2020-04-07)
Chelsea A Kraynak, Derek J Yan, Laura J Suggs
ABSTRACT
Macrophages play a critical role in the initiation, maintenance, and resolution of inflammation because of their diverse and plastic phenotypic responses to extracellular stimuli. Inflammatory stimuli drive the recruitment and activation of inflammatory (M1) macrophages, capable of significant cytokine production that potentiates inflammation. Local environmental signals including apoptotic cell efferocytosis drive a phenotypic transition toward pro-reparative (M2) macrophages to facilitate the resolution of inflammation. However, prolonged or dysregulated inflammatory macrophage response contributes to many disease states and tissue damage. We have developed a nanoparticle to help resolve macrophage-mediated inflammation by mimicking the anti-inflammatory effect of apoptotic cell engulfment. The nanoparticle, comprised of a poly(lactide-co-glycolide) core, is coated in phosphatidylserine (PS)-supplemented cell plasma membrane to emulate key characteristics of the apoptotic cell surface. These apoptotic body-inspired PS/membrane-coated nanoparticles (PS-MNPs) reduce inflammatory cytokine expression to promote an anti-inflammatory, phenotypic shift in macrophages in vitro, without the use of small molecule inhibitors or other drugs. Specifically, PS-MNP treatment before lipopolysaccharide (LPS)-induced inflammatory challenge resulted in a 2.5-fold reduction in secreted tumor necrosis factor α (TNFα) at 24 h, with co-treatment of PS-MNPs and LPS demonstrating a 5-fold TNFα reduction compared to LPS alone. Reduced TNFα production, as well as gene expression of several pro-inflammatory cytokines, correlated with a reduction in NFκB activation from PS-MNP treatment. The development of a nanoparticle to reduce the production of multiple inflammatory cytokines and transition away from an inflammatory macrophage phenotype, through the use of a physiologic anti-inflammatory pathway, illustrates a new potential strategy in creating anti-inflammatory therapeutics. STATEMENT OF SIGNIFICANCE: Macrophages propagate inflammation as the major source of cytokine production in the body. In inflammatory diseases, pro-inflammatory macrophages persist in the site of inflammation and exacerbate tissue destruction. Current anti-inflammatory drugs have significant drawbacks, including variable response rates and off-target effects. Here, we have developed an apoptotic-body inspired nanoparticle to modulate inflammatory macrophage phenotype. This polymeric nanoparticle is coated with phosphatidylserine-supplemented cell plasma membrane to mimic the anti-inflammatory effect of apoptotic cell engulfment. Nanoparticle delivery reduces inflammatory cytokine production and promotes an anti-inflammatory phenotypic macrophage shift. The capacity of these nanoparticles to help resolve macrophage-mediated inflammation may be a useful tool to study macrophage-apoptotic cell interactions, the role of macrophages in inflammatory diseases, and in the design of anti-inflammatory therapeutics.
Product Description
Avanti
16:0 PS, Avanti Research - A Croda Brand