Merck
  • Impact of Right Atrial Physiology on Heart Failure and Adverse Events after Myocardial Infarction.
Impact of Right Atrial Physiology on Heart Failure and Adverse Events after Myocardial Infarction.
Journal of clinical medicine (2020-01-17)
Andreas Schuster, Sören J Backhaus, Thomas Stiermaier, Jenny-Lou Navarra, Johannes Uhlig, Karl-Philipp Rommel, Alexander Koschalka, Johannes T Kowallick, Boris Bigalke, Shelby Kutty, Matthias Gutberlet, Gerd Hasenfuß, Holger Thiele, Ingo Eitel
ABSTRACT
Background: Right ventricular (RV) function is a known predictor of adverse events in heart failure and following acute myocardial infarction (AMI). While right atrial (RA) involvement is well characterized in pulmonary arterial hypertension, its relative contributions to adverse events following AMI especially in patients with heart failure and congestion need further evaluation. Methods: In this cardiovascular magnetic resonance (CMR)-substudy of AIDA STEMI and TATORT NSTEMI, 1235 AMI patients underwent CMR after primary percutaneous coronary intervention (PCI) in 15 centers across Germany (n = 795 with ST-elevation myocardial infarction and 440 with non-ST-elevation MI). Right atrial (RA) performance was evaluated using CMR myocardial feature tracking (CMR-FT) for the assessment of RA reservoir (total strain εs), conduit (passive strain εe), booster pump function (active strain εa), and associated strain rates (SR) in a blinded core-laboratory. The primary endpoint was the occurrence of major adverse cardiac events (MACE) 12 months post AMI. Results: RA reservoir (εsp = 0.061, SRs p = 0.049) and conduit functions (εep = 0.006, SRe p = 0.030) were impaired in patients with MACE as opposed to RA booster pump (εap = 0.579, SRa p = 0.118) and RA volume index (p = 0.866). RA conduit function was associated with the clinical onset of heart failure and MACE independently of RV systolic function and atrial fibrillation (AF) (multivariable analysis hazard ratio 0.95, 95% confidence interval 0.92 to 0.99, p = 0.009), while RV systolic function and AF were not independent prognosticators. Furthermore, RA conduit strain identified low- and high-risk groups within patients with reduced RV systolic function (p = 0.019 on log rank testing). Conclusions: RA impairment is a distinct feature and independent risk factor in patients following AMI and can be easily assessed using CMR-FT-derived quantification of RA strain.
Sigma-Aldrich
AIDA, solid
Pricing and availability is not currently available.