Merck
  • Characterisation and utilization of steel industry waste sludge as heterogeneous catalyst for the abatement of chlorinated organics by advanced oxidation processes.
Characterisation and utilization of steel industry waste sludge as heterogeneous catalyst for the abatement of chlorinated organics by advanced oxidation processes.
Chemosphere (2019-11-07)
Swati Singh, Anurag Garg
ABSTRACT
Catalytic advanced oxidation processes (AOPs) utilising UV irradiation have been reported to be highly efficient for the degradation of recalcitrant compounds. The focus of present study was to evaluate the potential of steel industry waste as an alternative to homogenous iron salts often used in AOPs. Iron rich sludge from effluent treatment plant (ETP) of a steel industry was subjected to acid washing and acid washing followed by calcination before using as catalyst for the oxidative removal of chlorinated organics present in synthetic and simulated bleaching effluents. The maximum total organic carbon removals of 64% and 25% from 4-chlorophenol solution and simulated pulp bleaching effluent, respectively were observed during photo-catalytic oxidation with acid washed steel sludge at stoichiometric H2O2 dose after 2 h reaction (catalyst dose = 1 g/L, and pH = 4.4). Low iron leaching (<2%) was observed from the catalyst even in acidic conditions and it could be reused twice without significant loss of catalytic activity.
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
Potassium titanium oxide oxalate dihydrate