Merck
Ras isoforms: signaling specificities in CD40 pathway.
Cell communication and signaling : CCS (2020-01-08)
Arathi Nair, Sushmita Chakraborty, Late Anirban Banerji, Ankita Srivastava, Charudutta Navare, Bhaskar Saha
ABSTRACT
Ras are small cellular GTPases which regulate diverse cellular processes. It has three isoforms: H-Ras, K-Ras, and N-Ras. Owing to the N-terminus (1-165 residues) sequence homology these isoforms were thought to be functionally redundant. However, only K-Ras-deficient mice but not H-Ras- and N-Ras-deficient mice show embryonic lethality. Similarly, mutations in a given Ras isoform are associated with a particular type of cancer. Moreover, we have previously reported that Ras isoforms perform unique functions in Leishmania major infection. Thus, Ras isoforms are implicated to have signaling and functional specificity but the mechanism remains to be elucidated. Using CD40 as a model receptor, we showed that depending on the strength of signaling, specific Ras isoforms are activated. Weak CD40 signal activates N-Ras, whereas strong signal activates H-Ras and K-Ras. Additionally, we showed that suppression of N-Ras expression reduced CD40-induced extracellular signal-regulated kinase-1/2 (ERK-1/2) activation and Interleukin (IL)-10 production; whereas suppression of H-Ras or K-Ras reduced CD40-induced p38 mitogen-activated protein kinase (p38MAPK) activation and IL-12 production. Furthermore, we showed that Ras isoforms have activator (GEF) specificity as weak CD40 signal-activated N-Ras requires Sos-1/2 whereas strong CD40 signal-activated H-Ras/K-Ras requires Ras-GRP as the guanine-nucleotide exchange factor (GEF) inducing ERK-1/2- or p38MAPK-mediated IL-10 or IL-12 productions, respectively, in macrophages. Silencing of syk reduced CD40-induced N-Ras activation but silencing of lyn inhibited H-Ras and K-Ras activation. In CD40 signaling, Ras isoforms also showed effector specificity; while H-Ras and K-Ras showed specificity for phosphatidyl inositol-3 kinase activation at high dose of CD40 stimulation, N-Ras primarily associated with Raf-1 at low dose of CD40 stimulation. Moreover, fractal analysis showed that functional site surface roughness for H-Ras (SurfaceFD = 2.39) and K-Ras (SurfaceFD = 2.39) are similar but significantly different from N-Ras (SurfaceFD = 2.25). The activator and effector specificities of Ras isoforms in CD40 signaling indicated their differential involvement in CD40 pathway and in maintaining the reciprocity. Our observations reveal Ras-regulated signaling outcome and its potential for developing Ras isoform-targeted immunotherapy and prophylaxis.
Sigma-Aldrich
MISSION® esiRNA, targeting human KRAS
Sigma-Aldrich
MISSION® esiRNA, targeting human NRAS
Sigma-Aldrich
MISSION® esiRNA, targeting human LYN
Sigma-Aldrich
MISSION® esiRNA, targeting human VAV1
Sigma-Aldrich
MISSION® esiRNA, targeting human HRAS