Merck
  • Visible-Light-Mediated Aerobic Tandem Dehydrogenative Povarov/Aromatization Reaction: Synthesis of Isocryptolepines.
Visible-Light-Mediated Aerobic Tandem Dehydrogenative Povarov/Aromatization Reaction: Synthesis of Isocryptolepines.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-09-26)
Eva Schendera, Lisa-Natascha Unkel, Phung Phan Huyen Quyen, Gwen Salkewitz, Frank Hoffmann, Alexander Villinger, Malte Brasholz
ABSTRACT
A metal-free, photoinduced aerobic tandem amine dehydrogenation/Povarov cyclization/aromatization reaction between N-aryl glycine esters and indoles leads to tetracyclic 11H-indolo[3,2-c]quinolines under mild conditions and with high yields. The reaction can be performed by using molecular iodine along with visible light, or by combining an organic photoredox catalyst with a halide anion. Mechanistic studies reveal that product formation occurs through a combination of radical-mediated oxidation steps with an iminium ion or N-haloiminium ion [4+2]-cycloaddition, and the N-heterocyclic products constitute new analogues of the antiplasmodial natural alkaloid isocryptolepine.