Merck
  • A fluorometric assay for glycosylasparaginase activity and detection of aspartylglycosaminuria.
A fluorometric assay for glycosylasparaginase activity and detection of aspartylglycosaminuria.
Analytical biochemistry (1993-02-01)
I T Mononen, V M Kaartinen, J C Williams
ABSTRACT
Recent experimental work on the mechanism of action of glycosylasparaginase suggests that the enzyme specifically reacts toward the L-asparagine or L-aspartic acid moiety of its substrates. Based on this, a new sensitive assay for glycosylasparaginase activity has been developed using L-aspartic acid beta-(7-amido-4-methylcoumarin) as substrate. Release of 7-amino-4-methylcoumarin was determined fluorometrically. At pH 7.5, Km = 93 microM, and as little as 1 ng of glycosylasparaginase could be detected with the assay. Hydrolysis of the substrate was inhibited by diazo-oxonorvaline, a specific inhibitor of glycosylasparaginase. In biological samples, the fluorometric assay is 40-100 times more sensitive than other published methods for glycosylasparaginase. This new assay enables a rapid enzymatic diagnosis of aspartylglycosaminuria--a genetic deficiency of glycosylasparaginase activity--with leukocyte and fibroblast samples.
Sigma-Aldrich
L-Aspartic acid β-(7-amido-4-methylcoumarin), fluorescent amino acid