- Tumor Necrosis Factor Receptor-Associated Factor 5 Promotes Arterial Neointima Formation through Smooth Muscle Cell Proliferation.
Tumor Necrosis Factor Receptor-Associated Factor 5 Promotes Arterial Neointima Formation through Smooth Muscle Cell Proliferation.
Tumor necrosis factor (TNF) receptor-associated factors (TRAFs) are cytoplasmic adaptor proteins of the TNF/interleukin (IL)-1/Toll-like receptor superfamily. Ligands of this family such as TNFα, CD40L, and IL-1β promote chronic inflammatory processes such as atherosclerosis and restenosis, the latter being a common adverse reaction after vascular interventions. We previously reported overexpression of TRAF5 in murine and human atheromata and TRAF5-dependent proinflammatory functions in vitro. However, the role of TRAF5 in restenosis remains unsettled. To evaluate whether TRAF5 affects neointima formation, TRAF5-/-LDLR-/- and TRAF5+/+LDLR-/- mice consuming a high cholesterol diet (HCD) received wire-induced injury of the carotid artery. After 28 days, TRAF5-deficient mice showed a 45% decrease in neointimal area formation compared with TRAF5-compentent mice. Furthermore, neointimal vascular smooth muscle cells (vSMC) and macrophages decreased whereas collagen increased in TRAF5-deficient mice. Mechanistically, the latter expressed lower transcript levels of the matrix metalloproteinases 2 and 9, both instrumental in extracellular matrix degradation and vSMC mobilization. Additionally, TRAF5-specific siRNA interference rendered murine vSMC less proliferative upon CD40L stimulation. In accordance with these findings, fewer vSMC isolated from TRAF5-deficient aortas were in a proliferative state as assessed by Ki67 and cyclin B1 expression. In conclusion, TRAF5 deficiency mitigates neointima formation in mice, likely through a TRAF5-dependent decrease in vSMC proliferation.