Merck
  • Development of Microalgae Biosensor Chip by Incorporating Microarray Oxygen Sensor for Pesticides Sensing.
Development of Microalgae Biosensor Chip by Incorporating Microarray Oxygen Sensor for Pesticides Sensing.
Biosensors (2019-11-16)
Md Abul Kashem, Kazuki Kimoto, Yasunori Iribe, Masayasu Suzuki
ABSTRACT
A microalgae (Pseudokirchneriella subcapitata) biosensor chip for pesticide sensing has been developed by attaching the immobilized microalgae biofilm pon the microarray dye spots (size 100 μm and pitch 200 μm). The dye spots (ruthenium complex) were printed upon SO3-modified glass slides using a polydimethylsiloxane (PDMS) stamp and a microcontact printer (μCP). Emitted fluorescence intensity (FI) variance due to photosynthetic activity (O2 production) of microalgae was monitored by an inverted fluorescent microscope and inhibition of the oxygen generation rate was calculated based on the FI responses both before and after injection of pesticide sample. The calibration curves, as the inhibition of oxygen generation rate (%) due to photosynthetic activity inhibition by the pesticides, depicted that among the 6 tested pesticides, the biosensor showed good sensitivity for 4 pesticides (diuron, simetryn, simazine, and atrazine) but was insensitive for mefenacet and pendimethalin. The detection limits were 1 ppb for diuron and 10 ppb for simetryn, simazine, and atrazine. The simple and low-cost nature of sensing of the developed biosensor sensor chip has apparently created opportunities for regular water quality monitoring, where pesticides are an important concern.
Product Description
Supelco
Simetryn, PESTANAL®, analytical standard