Merck
  • Transition Metal Coordination Polymers with Trans-1,4-Cyclohexanedicarboxylate: Acidity-Controlled Synthesis, Structures and Properties.
Transition Metal Coordination Polymers with Trans-1,4-Cyclohexanedicarboxylate: Acidity-Controlled Synthesis, Structures and Properties.
Materials (Basel, Switzerland) (2020-01-23)
Pavel A Demakov, Artem S Bogomyakov, Artem S Urlukov, Aleksandra Yu Andreeva, Denis G Samsonenko, Danil N Dybtsev, Vladimir P Fedin
ABSTRACT
Five trans-1,4-cyclohexanedicarboxylate (chdc2-) metal-organic frameworks of transition metals were synthesized in aqueous systems. A careful control of pH, reaction temperature and solvent composition were shown to direct the crystallization of a particular compound. Isostructural [Co(H2O)4(chdc)]n (1) and [Fe(H2O)4(chdc)]n (2) consist of one-dimensional hydrogen-bonded chains. Compounds [Cd(H2O)(chdc)]n∙0.5nCH3CN (3), [Mn4(H2O)3(chdc)4]n (4) and [Mn2(Hchdc)2(chdc)]n (5) possess three-dimensional framework structures. The compounds 1, 4 and 5 were further characterized by magnetochemical analysis, which reveals paramagnetic nature of these compounds. A presence of antiferromagnetic exchange at low temperatures is observed for 5 while the antiferromagnetic coupling in 4 is rather strong, even at ambient conditions. The thermal decompositions of 1, 4 and 5 were investigated and the obtained metal oxide (cubic Co3O4 and MnO) samples were analyzed by X-ray diffraction and scanning electron microscopy.