Merck
  • Generation of a PARK2 homozygous knockout induced pluripotent stem cell line (GIBHi002-A-1) with two common isoforms abolished.
Generation of a PARK2 homozygous knockout induced pluripotent stem cell line (GIBHi002-A-1) with two common isoforms abolished.
Stem cell research (2019-11-08)
Meng Zhang, David P Ibañez, Wenxia Fan, Hao Liu, Xiaofen Zhong, Xiwei Wang, Yingying Li, Mazid Md Abdul, Wenjuan Li, Yunpan Li, Carl Ward, Shuhan Chen, Dongye Wang, Baoming Qin, Miguel A Esteban, Ping Zhao, Zhiwei Luo
ABSTRACT
Loss of function mutations in PARK2 (encoding PARKIN) cause autosomal recessive Parkinson's disease (PD), which often manifests at a juvenile age. Molecular and biochemical studies show that PARKIN functions as an E3 ubiquitin ligase controlling mitochondrial homeostasis. Yet, the exact mechanisms are unclear due to the use of sub-optimal models including cancer cells and fibroblasts. We have generated a PARK2 knockout (KO) isogenic cell line using a well-characterized induced pluripotent stem cell (iPSC) clone with good differentiation potential. This cell line lacks the expression of all PARKIN isoforms and is valuable for elucidating the role of PARK2 mutations in PD.
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Lysis solution for blood, sufficient for 100 reactions, Molecular Biology
Sigma-Aldrich
CryoStor® cell cryopreservation media, CS10