Merck
  • Fgf10-Hippo Epithelial-Mesenchymal Crosstalk Maintains and Recruits Lung Basal Stem Cells.
Fgf10-Hippo Epithelial-Mesenchymal Crosstalk Maintains and Recruits Lung Basal Stem Cells.
Developmental cell (2017-10-11)
Thomas Volckaert, Tingting Yuan, Cho-Ming Chao, Harold Bell, Alina Sitaula, Luisa Szimmtenings, Elie El Agha, Diptiman Chanda, Susan Majka, Saverio Bellusci, Victor J Thannickal, Reinhard Fässler, Stijn P De Langhe
ABSTRACT
The lung harbors its basal stem/progenitor cells (BSCs) in the protected environment of the cartilaginous airways. After major lung injuries, BSCs are activated and recruited to sites of injury. Here, we show that during homeostasis, BSCs in cartilaginous airways maintain their stem cell state by downregulating the Hippo pathway (resulting in increased nuclear Yap), which generates a localized Fgf10-expressing stromal niche; in contrast, differentiated epithelial cells in non-cartilaginous airways maintain quiescence by activating the Hippo pathway and inhibiting Fgf10 expression in airway smooth muscle cells (ASMCs). However, upon injury, surviving differentiated epithelial cells spread to maintain barrier function and recruit integrin-linked kinase to adhesion sites, which leads to Merlin degradation, downregulation of the Hippo pathway, nuclear Yap translocation, and expression and secretion of Wnt7b. Epithelial-derived Wnt7b, then in turn, induces Fgf10 expression in ASMCs, which extends the BSC niche to promote regeneration.
Sigma-Aldrich
Anti-FGF10 Antibody, from rabbit, purified by affinity chromatography
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep