Merck
  • The farnesyltransferase inhibitor R115777 (ZARNESTRA) enhances the pro-apoptotic activity of interferon-alpha through the inhibition of multiple survival pathways.
The farnesyltransferase inhibitor R115777 (ZARNESTRA) enhances the pro-apoptotic activity of interferon-alpha through the inhibition of multiple survival pathways.
International journal of cancer (2007-07-28)
Michele Caraglia, Monica Marra, Caterina Viscomi, Anna Maria D'Alessandro, Alfredo Budillon, Giuseppina Meo, Claudio Arra, Antonio Barbieri, Ulf R Rapp, Alfonso Baldi, Pierfrancesco Tassone, Salvatore Venuta, Alberto Abbruzzese, Pierosandro Tagliaferri
ABSTRACT
Interferon alpha (IFNalpha) induces an EGF-Ras-->Raf-1-->Erk dependent survival pathway counteracting apoptosis induced by the cytokine. In this paper we have evaluated the effects of the combination between farnesyl-transferase inhibitor (FTI) R115777 and IFNalpha on the growth inhibition and apoptosis of cancer cells. Simultaneous exposure to R115777 and IFNalpha produced synergistic both antiproliferative and proapoptotic effects. In these experimental conditions, IFNalpha and R115777 completely antagonized the increased activity of both Ras and Erk-1/2 induced by IFNalpha and strongly reduced Akt activity. Furthermore, treatment with R115777 in combination with IFNalpha regimen induced tumor growth delay on established KB cell xenografts in nude mice, while the single agents were almost inactive. R115777 was again able to antagonize the Ras-dependent survival pathway induced by IFNalpha also in vivo. Raf-1, one of the downstream targets of Ras, has been reported to activate bcl-2 through displacement and/or phosphorylation of Bad. We have found that IFNalpha induced mitochondrial localization of Raf-1 that was antagonized by R115777. Moreover, IFNalpha increased Raf-1/bcl-2 immuno-conjugate formation and intracellular co-localization and enhanced phosphorylation of Bad at Ser 112 and again R115777 counteracted all these effects. Moreover, the use of plasmids encoding for dominant negative or dominant positive Raf-1 antagonized and potentiated, respectively, the co-immunoprecipitation between Raf-1 and bcl-2. In conclusion, FTI R115777 strongly potentiates the antitumor activity of IFNalpha both in vitro and in vivo through the inhibition of different survival pathways that are dependent from isoprenylation of intracellular proteins such as ras.
Sigma-Aldrich
Anti-Pan-Ras Antibody, clone RAS 10, clone RAS 10, from mouse
Pricing and availability is not currently available.