Merck
  • Nutrient modification of the innate immune response: a novel mechanism by which saturated fatty acids greatly amplify monocyte inflammation.
Nutrient modification of the innate immune response: a novel mechanism by which saturated fatty acids greatly amplify monocyte inflammation.
Arteriosclerosis, thrombosis, and vascular biology (2010-01-30)
Eric A Schwartz, Wei-Yang Zhang, Sheetal K Karnik, Sabine Borwege, Vijay R Anand, Phyllis S Laine, Yali Su, Peter D Reaven
ABSTRACT
Monocyte/macrophage inflammation is an important contributor to diabetes and cardiovascular disease. Studies have suggested saturated fatty acids (SFA) induce monocyte inflammation in a Toll-like receptor-4-dependent manner, but recent data suggest SFA do not directly interact with Toll-like receptor-4. The present study tests the novel hypothesis that metabolism of SFA cooperatively amplifies Toll-like receptor-4-mediated inflammation. THP-1 monocytes exposed to 100 micromol/L SFA in vitro for 16 hours followed by 1 ng/mL lipopolysaccharide demonstrated enhanced IL-6 and IL-8 mRNA and protein expression (approximately 3-fold higher than the sum of individual responses to SFA and lipopolysaccharide). SFA had similar effects on THP-1 macrophages and primary human monocytes. This amplified lipopolysaccharide response could be blocked by inhibition of SFA metabolism to ceramide and restored by cell-permeable ceramide. Both SFA and ceramide activated PKC-zeta and the mitogen-activated protein kinases Erk, JNK, and p38. Inhibition of these pathways prevented the SFA-induced increase in cytokine expression. These results provide evidence for potent amplification of monocyte/macrophage innate immune responses by a novel pathway requiring metabolism of SFA to ceramide and activation of PKC-zeta/mitogen-activated protein kinases. These findings demonstrate how nutrient excess may modulate innate immune system activation and possibly contribute to development of diabetes and cardiovascular disease.
Sigma-Aldrich
Anti-NFκB Antibody, p65 subunit, active subunit, clone 12H11, clone 12H11, Chemicon®, from mouse