Merck
  • Editing an α-globin enhancer in primary human hematopoietic stem cells as a treatment for β-thalassemia.
Editing an α-globin enhancer in primary human hematopoietic stem cells as a treatment for β-thalassemia.
Nature communications (2017-09-06)
Sachith Mettananda, Chris A Fisher, Deborah Hay, Mohsin Badat, Lynn Quek, Kevin Clark, Philip Hublitz, Damien Downes, Jon Kerry, Matthew Gosden, Jelena Telenius, Jackie A Sloane-Stanley, Paula Faustino, Andreia Coelho, Jessica Doondeea, Batchimeg Usukhbayar, Paul Sopp, Jacqueline A Sharpe, Jim R Hughes, Paresh Vyas, Richard J Gibbons, Douglas R Higgs
ABSTRACT
β-Thalassemia is one of the most common inherited anemias, with no effective cure for most patients. The pathophysiology reflects an imbalance between α- and β-globin chains with an excess of free α-globin chains causing ineffective erythropoiesis and hemolysis. When α-thalassemia is co-inherited with β-thalassemia, excess free α-globin chains are reduced significantly ameliorating the clinical severity. Here we demonstrate the use of CRISPR/Cas9 genome editing of primary human hematopoietic stem/progenitor (CD34+) cells to emulate a natural mutation, which deletes the MCS-R2 α-globin enhancer and causes α-thalassemia. When edited CD34+ cells are differentiated into erythroid cells, we observe the expected reduction in α-globin expression and a correction of the pathologic globin chain imbalance in cells from patients with β-thalassemia. Xenograft assays show that a proportion of the edited CD34+ cells are long-term repopulating hematopoietic stem cells, demonstrating the potential of this approach for translation into a therapy for β-thalassemia.β-thalassemia is characterised by the presence of an excess of α-globin chains, which contribute to erythrocyte pathology. Here the authors use CRISP/Cas9 to reduce α-globin expression in hematopoietic precursors, and show effectiveness in xenograft assays in mice.
Sigma-Aldrich
Chromatin Immunoprecipitation (ChIP) Assay Kit, Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads.
Sigma-Aldrich
ChIPAb+ Acetyl-Histone H4 - ChIP Validated Antibody and Primer Set, serum, from rabbit