Merck
  • Conversion of human umbilical cord mesenchymal stem cells in Wharton's jelly to dopaminergic neurons in vitro: potential therapeutic application for Parkinsonism.
Conversion of human umbilical cord mesenchymal stem cells in Wharton's jelly to dopaminergic neurons in vitro: potential therapeutic application for Parkinsonism.
Stem cells (Dayton, Ohio) (2005-08-16)
Yu-Show Fu, Yun-Chih Cheng, Maan-Yuh Anya Lin, Henrich Cheng, Pei-Ming Chu, Shih-Chich Chou, Yang-Hsin Shih, Miau-Hwa Ko, Min-Shan Sung
ABSTRACT
Human mesenchymal stem cells isolated from Wharton's jelly of the umbilical cord were induced to transform into dopaminergic neurons in vitro through stepwise culturing in neuron-conditioned medium, sonic hedgehog, and FGF8. The success rate was 12.7%, as characterized by positive staining for tyrosine hydroxylase (TH), the rate-limiting catecholaminergic synthesizing enzyme, and dopamine being released into the culture medium. Transplantation of such cells into the striatum of rats previously made Parkinsonian by unilateral striatal lesioning with the dopaminergic neurotoxin 6-hydroxydopamine partially corrected the lesion-induced amphetamine-evoked rotation. Viability of the transplanted cells at least 4 months after transplantation was identified by positive TH staining and migration of 1.4 mm both rostrally and caudally. These results suggest that human umbilical mesenchymal stem cells have the potential for treatment of Parkinson's disease.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Rhodamine conjugate, Chemicon®, from goat
Sigma-Aldrich
Anti-Dopamine β Hydroxylase Antibody, clone 4F10.2, ascites fluid, clone 4F10.2, Chemicon®
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) FITC Conjugated, 2 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Anti-Nuclei Antibody, clone 235-1, clone 235-1, Chemicon®, from mouse