Merck
  • Combinatorial Targeting by MicroRNAs Co-ordinates Post-transcriptional Control of EMT.
Combinatorial Targeting by MicroRNAs Co-ordinates Post-transcriptional Control of EMT.
Cell systems (2018-07-17)
Joseph Cursons, Katherine A Pillman, Kaitlin G Scheer, Philip A Gregory, Momeneh Foroutan, Soroor Hediyeh-Zadeh, John Toubia, Edmund J Crampin, Gregory J Goodall, Cameron P Bracken, Melissa J Davis
ABSTRACT
MicroRNAs (miRNAs) are important post-transcriptional regulators of gene expression, functioning in part by facilitating the degradation of target mRNAs. They have an established role in controlling epithelial-mesenchymal transition (EMT), a reversible phenotypic program underlying normal and pathological processes. Many studies demonstrate the role of individual miRNAs using overexpression at levels greatly exceeding physiological abundance. This can influence transcripts with relatively poor targeting and may in part explain why over 130 different miRNAs are directly implicated as EMT regulators. Analyzing a human mammary cell model of EMT we found evidence that a set of miRNAs, including the miR-200 and miR-182/183 family members, co-operate in post-transcriptional regulation, both reinforcing and buffering transcriptional output. Investigating this, we demonstrate that combinatorial treatment altered cellular phenotype with miRNA concentrations much closer to endogenous levels and with less off-target effects. This suggests that co-operative targeting by miRNAs is important for their physiological function and future work classifying miRNAs should consider such combinatorial effects.
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-MMP-1 Antibody, hinge region, from rabbit, purified by affinity chromatography
Sigma-Aldrich
ChIPAb+ Acetyl-Histone H3 (Lys27) - ChIP Validated Antibody and Primer Set, clone CMA309, from mouse