- CB1 positive allosteric modulation attenuates Δ9-THC withdrawal and NSAID-induced gastric inflammation.
CB1 positive allosteric modulation attenuates Δ9-THC withdrawal and NSAID-induced gastric inflammation.
Recently, multiple compounds have been synthesized that target the allosteric binding site(s) of CB1. These CB1 positive allosteric modulators may capture the benefits of cannabinoid receptor activation without unwanted psychoactive effects, such as sedation. For example, ZCZ011 blocks neuropathic pain, absent the catalepsy, sedation, and hypothermia caused by CB1 orthosteric modulators, including Δ9-tetrahydrocannabinol (THC). The primary goal of the present study was to evaluate the potential of ZCZ011 to attenuate somatic signs of cannabinoid withdrawal in mice. Mice were repeatedly administered THC (10 mg/kg, s.c.) or vehicle, and withdrawal was either precipitated using the CB1 antagonist rimonabant (3 mg/kg, i.p.) or elicited spontaneously via THC abstinence. ZCZ011 (≥10 mg/kg, i.p.) significantly attenuated somatic signs of withdrawal, including head twitches and paw tremors, but had no effect on locomotor activity or conditioned place preference. We next tested the antiulcerogenic properties of CB1 positive allosteric modulation. Mice were fasted for 22 h, administered ZCZ011, and gastric hemorrhages were induced with the nonsteroidal anti-inflammatory drug diclofenac sodium (100 mg/kg, p.o.). ZCZ011 alone had no effect on gastric ulceration, but ZCZ011 (≥10 mg/kg) blocked ulcer formation when combined with a subthreshold MAGL inhibitor (JZL184; 1 mg/kg, i.p.). Thus, CB1 positive allosteric modulation is a novel approach to treat cannabinoid dependence and gastric inflammation.