Merck
  • An efficient CRISPR-based strategy to insert small and large fragments of DNA using short homology arms.
An efficient CRISPR-based strategy to insert small and large fragments of DNA using short homology arms.
eLife (2019-11-02)
Oguz Kanca, Jonathan Zirin, Jorge Garcia-Marques, Shannon Marie Knight, Donghui Yang-Zhou, Gabriel Amador, Hyunglok Chung, Zhongyuan Zuo, Liwen Ma, Yuchun He, Wen-Wen Lin, Ying Fang, Ming Ge, Shinya Yamamoto, Karen L Schulze, Yanhui Hu, Allan C Spradling, Stephanie E Mohr, Norbert Perrimon, Hugo J Bellen
ABSTRACT
We previously reported a CRISPR-mediated knock-in strategy into introns of Drosophila genes, generating an attP-FRT-SA-T2A-GAL4-polyA-3XP3-EGFP-FRT-attP transgenic library for multiple uses (Lee et al., 2018a). The method relied on double stranded DNA (dsDNA) homology donors with ~1 kb homology arms. Here, we describe three new simpler ways to edit genes in flies. We create single stranded DNA (ssDNA) donors using PCR and add 100 nt of homology on each side of an integration cassette, followed by enzymatic removal of one strand. Using this method, we generated GFP-tagged proteins that mark organelles in S2 cells. We then describe two dsDNA methods using cheap synthesized donors flanked by 100 nt homology arms and gRNA target sites cloned into a plasmid. Upon injection, donor DNA (1 to 5 kb) is released from the plasmid by Cas9. The cassette integrates efficiently and precisely in vivo. The approach is fast, cheap, and scalable.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-ACOX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2