Merck
  • Lysine demethylase 2 (KDM2B) regulates hippo pathway via MOB1 to promote pancreatic ductal adenocarcinoma (PDAC) progression.
Lysine demethylase 2 (KDM2B) regulates hippo pathway via MOB1 to promote pancreatic ductal adenocarcinoma (PDAC) progression.
Journal of experimental & clinical cancer research : CR (2020-01-17)
Ming Quan, Zhiqin Chen, Feng Jiao, Xiuying Xiao, Qing Xia, Jingde Chen, Qian Chao, Yandong Li, Yong Gao, Haiyan Yang, Liwei Wang, Jiujie Cui
ABSTRACT
Mps1 binding protein (MOB1) is one of the core components of the mammalian Hippo pathway and plays important roles in cancer development. However, its expression, function and regulation in pancreatic ductal adenocarcinoma (PDAC) have not been revealed yet. The expression of MOB1 and lysine demethylase 2B (KDM2B) in PDAC and adjacent normal pancreas tissues were measured. Also, the underlying mechanisms of altered MOB1 expression and its impact on PDAC biology were investigated. We revealed for the first time that MOB1 was decreased expression in PDAC and was a statistically significant independent predictor of poor survival, and restored expression of MOB1 suppressed the proliferation, migration and invasion of PDAC cells. Further studies demonstrated that KDM2B directly bound to the promoter region of MOB1, and suppressed the promoter activity of MOB1 and transcriptionally inhibited the MOB1 expression. Furthermore, KDM2B regulated Hippo pathway and promoted PDAC proliferation, migration and invasion via MOB1. This study demonstrated the mechanism and roles of a novel KDM2B/MOB1/Hippo signaling in PDAC progression.
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-JHDM1B Antibody, from rabbit, purified by affinity chromatography
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-MOB1A antibody produced in rabbit, affinity isolated antibody