Merck
  • Fasting Activates Fatty Acid Oxidation to Enhance Intestinal Stem Cell Function during Homeostasis and Aging.
Fasting Activates Fatty Acid Oxidation to Enhance Intestinal Stem Cell Function during Homeostasis and Aging.
Cell stem cell (2018-05-05)
Maria M Mihaylova, Chia-Wei Cheng, Amanda Q Cao, Surya Tripathi, Miyeko D Mana, Khristian E Bauer-Rowe, Monther Abu-Remaileh, Laura Clavain, Aysegul Erdemir, Caroline A Lewis, Elizaveta Freinkman, Audrey S Dickey, Albert R La Spada, Yanmei Huang, George W Bell, Vikram Deshpande, Peter Carmeliet, Pekka Katajisto, David M Sabatini, Ömer H Yilmaz
ABSTRACT
Diet has a profound effect on tissue regeneration in diverse organisms, and low caloric states such as intermittent fasting have beneficial effects on organismal health and age-associated loss of tissue function. The role of adult stem and progenitor cells in responding to short-term fasting and whether such responses improve regeneration are not well studied. Here we show that a 24 hr fast augments intestinal stem cell (ISC) function in young and aged mice by inducing a fatty acid oxidation (FAO) program and that pharmacological activation of this program mimics many effects of fasting. Acute genetic disruption of Cpt1a, the rate-limiting enzyme in FAO, abrogates ISC-enhancing effects of fasting, but long-term Cpt1a deletion decreases ISC numbers and function, implicating a role for FAO in ISC maintenance. These findings highlight a role for FAO in mediating pro-regenerative effects of fasting in intestinal biology, and they may represent a viable strategy for enhancing intestinal regeneration.
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture