Merck
  • A lineage-specific rapid diagnostic test (Chagas Sero K-SeT) identifies Brazilian Trypanosoma cruzi II/V/VI reservoir hosts among diverse mammalian orders.
A lineage-specific rapid diagnostic test (Chagas Sero K-SeT) identifies Brazilian Trypanosoma cruzi II/V/VI reservoir hosts among diverse mammalian orders.
PloS one (2020-01-18)
Mairi C W McClean, Tapan Bhattacharyya, Pascal Mertens, Niamh Murphy, Quentin Gilleman, Yves Gustin, Nicolas Zeippen, Samanta C C Xavier, Ana M Jansen, Michael A Miles
ABSTRACT
Trypanosoma cruzi, the protozoan agent of Chagas disease in the Americas, is comprised of six genetic lineages (TcI-TcVI) and a possible seventh (TcBat, related to TcI). Identification of T. cruzi lineages infecting reservoir mammalian species is fundamental to resolving transmission cycles. However, this is hindered by the limited sensitivity and technical complexity of parasite isolation and genotyping. An alternative approach is serology using T. cruzi lineage-specific epitopes, such as those of the trypomastigote small surface antigen (TSSA). For surveillance of T. cruzi lineage infections in mammal species from diverse Brazilian regions, we apply a novel rapid diagnostic test (RDT, Chagas Sero K-SeT), which incorporates the TSSA peptide epitope specific to TcII/V/VI (TSSApep-II/V/VI) and Protein G detection of antibodies. Chagas Sero K-SeT RDT results with sera from experimentally infected mice, from tamarin primates (Leontopithecus spp.) and from canines (Canis familiaris) were concordant with corresponding TSSApep-II/V/VI ELISAs. The Chagas Sero K-Set detected TcII/V/VI infections in Leontopithecus spp. from the Atlantic forest (n = 46), in C. familiaris (n = 16) and Thrichomys laurentius (n = 2) from Caatinga biome and Chiroptera (n = 1) from Acre, Amazonia. The Chagas Sero K-SeT RDT is directly applicable to TcII/V/VI-specific serological surveillance of T. cruzi infection in several different mammalian Orders. It can replace ELISAs and provides efficient, point-of-sampling, low-cost detection of TcII/V/VI infections, with at least equivalent sensitivity, although some mammals may be difficult to trap, and, not unexpectedly, Chagas Sero K-SeT could not recognise feline IgG. Knowledge of sylvatic hosts of T. cruzi can be expanded, new reservoir species discovered, and the ecology of transmission cycles clarified, particularly with adaptation to further mammalian Orders.
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Protein G−Peroxidase from Streptococcus sp., recombinant, expressed in unspecified host, lyophilized powder
Sigma-Aldrich
Anti-Cat IgG (H+L)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder