Merck
  • A novel environment-evoked transcriptional signature predicts reactivity in single dentate granule neurons.
A novel environment-evoked transcriptional signature predicts reactivity in single dentate granule neurons.
Nature communications (2018-08-08)
Baptiste N Jaeger, Sara B Linker, Sarah L Parylak, Jerika J Barron, Iryna S Gallina, Christian D Saavedra, Conor Fitzpatrick, Christina K Lim, Simon T Schafer, Benjamin Lacar, Sebastian Jessberger, Fred H Gage
ABSTRACT
Activity-induced remodeling of neuronal circuits is critical for memory formation. This process relies in part on transcription, but neither the rate of activity nor baseline transcription is equal across neuronal cell types. In this study, we isolated mouse hippocampal populations with different activity levels and used single nucleus RNA-seq to compare their transcriptional responses to activation. One hour after novel environment exposure, sparsely active dentate granule (DG) neurons had a much stronger transcriptional response compared to more highly active CA1 pyramidal cells and vasoactive intestinal polypeptide (VIP) interneurons. Activity continued to impact transcription in DG neurons up to 5 h, with increased heterogeneity. By re-exposing the mice to the same environment, we identified a unique transcriptional signature that selects DG neurons for reactivation upon re-exposure to the same environment. These results link transcriptional heterogeneity to functional heterogeneity and identify a transcriptional correlate of memory encoding in individual DG neurons.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, Alexa Fluor488 conjugated, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Prox1 Antibody, clone 4G10, clone 4G10, Chemicon®, from mouse