Merck
  • Anterior Gradient 3 Promotes Breast Cancer Development and Chemotherapy Response.
Anterior Gradient 3 Promotes Breast Cancer Development and Chemotherapy Response.
Cancer research and treatment : official journal of Korean Cancer Association (2019-07-12)
Qiao Xu, Ying Shao, Jinman Zhang, Huikun Zhang, Yawen Zhao, Xiaoli Liu, Zhifang Guo, Wei Chong, Feng Gu, Yongjie Ma
ABSTRACT
Anterior gradient 3 (AGR3) belongs to human anterior gradient (AGR) family. The function of AGR3 on cancer remains unknown. This research aimed to investigate if AGR3 had prognostic values in invasive ductal carcinoma (IDC) of breast cancer and could promote tumor progression. AGR3 expression was detected in breast benign lesions, ductal carcinoma in situ and IDC by immunohistochemistry analysis. AGR3's correlations with clinicopathological features and prognosis of IDC patients were analyzed. By cell function experiments, collagen gel droplet-embedded culture drug sensitivity test and cytotoxic analysis, AGR3's impacts on proliferation, invasion ability, and chemotherapeutic drug sensitivity of breast cancer cells were also detected. AGR3 was up-regulated in luminal subtype of histological grade I-II of IDC patients and positively correlated with high risks of recurrence and distant metastasis. AGR3 high expression could lead to bone or liver metastasis and predict poor prognosis of luminal B. In cell lines, AGR3 could promote proliferation and invasion ability of breast cancer cells which were consistent with clinical analysis. Besides, AGR3 could indicate poor prognosis of breast cancer patients treated with taxane but a favorable prognosis with 5-fluoropyrimidines. And breast cancer cells with AGR3 high expression were resistant to taxane but sensitive to 5-fluoropyrimidines. AGR3 might be a potential prognostic indicator in luminal B subtype of IDC patients of histological grade I-II. And patients with AGR3 high expression should be treated with chemotherapy regimens consisting of 5-fluoropyrimidines but no taxane.
Sigma-Aldrich
Anti-AGR3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution