Merck
  • 14-3-3ε and ζ regulate neurogenesis and differentiation of neuronal progenitor cells in the developing brain.
14-3-3ε and ζ regulate neurogenesis and differentiation of neuronal progenitor cells in the developing brain.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-09-05)
Kazuhito Toyo-oka, Tomoka Wachi, Robert F Hunt, Scott C Baraban, Shinichiro Taya, Hayley Ramshaw, Kozo Kaibuchi, Quenten P Schwarz, Angel F Lopez, Anthony Wynshaw-Boris
ABSTRACT
During brain development, neural progenitor cells proliferate and differentiate into neural precursors. These neural precursors migrate along the radial glial processes and localize at their final destination in the cortex. Numerous reports have revealed that 14-3-3 proteins are involved in many neuronal activities, although their functions in neurogenesis remain unclear. Here, using 14-3-3ε/ζ double knock-out mice, we found that 14-3-3 proteins are important for proliferation and differentiation of neural progenitor cells in the cortex, resulting in neuronal migration defects and seizures. 14-3-3 deficiency resulted in the increase of δ-catenin and the decrease of β-catenin and αN-catenin. 14-3-3 proteins regulated neuronal differentiation into neurons via direct interactions with phosphorylated δ-catenin to promote F-actin formation through a catenin/Rho GTPase/Limk1/cofilin signaling pathway. Conversely, neuronal migration defects seen in the double knock-out mice were restored by phosphomimic Ndel1 mutants, but not δ-catenin. Our findings provide new evidence that 14-3-3 proteins play important roles in neurogenesis and neuronal migration via the regulation of distinct signaling cascades.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-LIMK1 Antibody, clone 40/00-1B11-1-1, clone 40/00-1B11-1-1, from rat
Sigma-Aldrich
Anti-Tbr2 Antibody, from chicken, purified by affinity chromatography