Merck
  • CLPP deficiency protects against metabolic syndrome but hinders adaptive thermogenesis.
CLPP deficiency protects against metabolic syndrome but hinders adaptive thermogenesis.
EMBO reports (2018-03-29)
Christina Becker, Alexandra Kukat, Karolina Szczepanowska, Steffen Hermans, Katharina Senft, Christoph Paul Brandscheid, Priyanka Maiti, Aleksandra Trifunovic
ABSTRACT
Mitochondria are fundamental for cellular metabolism as they are both a source and a target of nutrient intermediates originating from converging metabolic pathways, and their role in the regulation of systemic metabolism is increasingly recognized. Thus, maintenance of mitochondrial homeostasis is indispensable for a functional energy metabolism of the whole organism. Here, we report that loss of the mitochondrial matrix protease CLPP results in a lean phenotype with improved glucose homeostasis. Whole-body CLPP-deficient mice are protected from diet-induced obesity and insulin resistance, which was not present in mouse models with either liver- or muscle-specific depletion of CLPP However, CLPP ablation also leads to a decline in brown adipocytes function leaving mice unable to cope with a cold-induced stress due to non-functional adaptive thermogenesis. These results demonstrate a critical role for CLPP in different metabolic stress conditions such as high-fat diet feeding and cold exposure providing tools to understand pathologies with deregulated Clpp expression and novel insights into therapeutic approaches against metabolic dysfunctions linked to mitochondrial diseases.
Sigma-Aldrich
Anti-GLUT-4 Antibody, C-terminus, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-CLPX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution