Merck
Sam68 Allows Selective Targeting of Human Cancer Stem Cells.
Cell chemical biology (2017-06-27)
Yannick D Benoit, Ryan R Mitchell, Ruth M Risueño, Luca Orlando, Borko Tanasijevic, Allison L Boyd, Lili Aslostovar, Kyle R Salci, Zoya Shapovalova, Jennifer Russell, Masakatsu Eguchi, Diana Golubeva, Monica Graham, Anargyros Xenocostas, Michael R Trus, Ronan Foley, Brian Leber, Tony J Collins, Mickie Bhatia
ABSTRACT
Targeting of human cancer stem cells (CSCs) requires the identification of vulnerabilities unique to CSCs versus healthy resident stem cells (SCs). Unfortunately, dysregulated pathways that support transformed CSCs, such as Wnt/β-catenin signaling, are also critical regulators of healthy SCs. Using the ICG-001 and CWP family of small molecules, we reveal Sam68 as a previously unappreciated modulator of Wnt/β-catenin signaling within CSCs. Disruption of CBP-β-catenin interaction via ICG-001/CWP induces the formation of a Sam68-CBP complex in CSCs that alters Wnt signaling toward apoptosis and differentiation induction. Our study identifies Sam68 as a regulator of human CSC vulnerability.
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
ChIPAb+ TCF-4 - ChIP Validated Antibody and Primer Set, from mouse
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys14) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-Sam 68 Antibody, serum, Upstate®