Merck
  • A retinoic acid-enhanced, multicellular human blood-brain barrier model derived from stem cell sources.
A retinoic acid-enhanced, multicellular human blood-brain barrier model derived from stem cell sources.
Scientific reports (2014-02-25)
Ethan S Lippmann, Abraham Al-Ahmad, Samira M Azarin, Sean P Palecek, Eric V Shusta
ABSTRACT
Blood-brain barrier (BBB) models are often used to investigate BBB function and screen brain-penetrating therapeutics, but it has been difficult to construct a human model that possesses an optimal BBB phenotype and is readily scalable. To address this challenge, we developed a human in vitro BBB model comprising brain microvascular endothelial cells (BMECs), pericytes, astrocytes and neurons derived from renewable cell sources. First, retinoic acid (RA) was used to substantially enhance BBB phenotypes in human pluripotent stem cell (hPSC)-derived BMECs, particularly through adherens junction, tight junction, and multidrug resistance protein regulation. RA-treated hPSC-derived BMECs were subsequently co-cultured with primary human brain pericytes and human astrocytes and neurons derived from human neural progenitor cells (NPCs) to yield a fully human BBB model that possessed significant tightness as measured by transendothelial electrical resistance (~5,000 Ωxcm(2)). Overall, this scalable human BBB model may enable a wide range of neuroscience studies.
Sigma-Aldrich
MK-571 sodium salt hydrate, ≥95% (HPLC)