- Toxicity and toxins of natural blooms and isolated strains of Microcystis spp. (Cyanobacteria) and improved procedure for purification of cultures.
Toxicity and toxins of natural blooms and isolated strains of Microcystis spp. (Cyanobacteria) and improved procedure for purification of cultures.
All samples of cyanobacterial blooms collected from 1986 to 1989 from Lake Kasumigaura, Ibaraki Prefecture, Japan, were hepatotoxic. The 50% lethal doses (LD50s) of the blooms to mice ranged from 76 to 556 mg/kg of body weight. Sixty-eight Microcystis cell clones (67 Microcystis aeruginosa and 1 M. viridis) were isolated from the blooms. Twenty-three strains (including the M. viridis strain) were toxic. However, the ratio of toxic to nontoxic strains among the blooms varied (6 to 86%). Microcystins were examined in six toxic strains. Five toxic strains produced microcystin-RR, -YR, and -LR, with RR being the dominant toxin in these strains. Another strain produced 7-desmethylmicrocystin-LR and an unknown microcystin. This strain showed the highest toxicity. Establishment of axenic strains from the Microcystis cells exhibiting extracellularly mucilaginous materials was successful by using a combination of the agar plate technique and two-step centrifugation.