Merck
  • Pharmacological profile of vascular activity of human stem villous arteries.
Pharmacological profile of vascular activity of human stem villous arteries.
Placenta (2019-10-02)
Katrin N Sander, Tayyba Y Ali, Averil Y Warren, Daniel P Hay, Fiona Broughton Pipkin, David A Barrett, Raheela N Khan
ABSTRACT
The function of the placental vasculature differs considerably from other systemic vascular beds of the human body. A detailed understanding of the normal placental vascular physiology is the foundation to understand perturbed conditions potentially leading to placental dysfunction. Behaviour of human stem villous arteries isolated from placentae at term pregnancy was assessed using wire myography. Effects of a selection of known vasoconstrictors and vasodilators of the systemic vasculature were assessed. The morphology of stem villous arteries was examined using IHC and TEM. Contractile effects in stem villous arteries were caused by U46619, 5-HT, angiotensin II and endothelin-1 (p ≤ 0.05), whereas noradrenaline and AVP failed to result in a contraction. Dilating effects were seen for histamine, riluzole, nifedipine, papaverine, SNP and SQ29548 (p ≤ 0.05) but not for acetylcholine, bradykinin and substance P. Stem villous arteries behave differently to vessels of the systemic vasculature and results indicate that the placenta is cut off from the systemic maternal vascular regulation. Particularly, endothelium-dependent processes were attenuated in the placental vasculature, creating a need to determine the role of the endothelium in the placenta in future studies.
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
Bradykinin acetate salt, powder, ≥98% (HPLC)
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder
Sigma-Aldrich
Angiotensin II human, ≥93% (HPLC), powder
Sigma-Aldrich
L-Norepinephrine hydrochloride, ≥98.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
[Arg8]-Vasopressin acetate salt, ≥95% (HPLC)
Sigma-Aldrich
Riluzole, solid
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Papaverine hydrochloride, powder
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC)
Sigma-Aldrich
Substance P acetate salt hydrate, ≥95% (HPLC), powder
Supelco
Serotonin, analytical standard
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC), free-flowing, Redi-Dri
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder