Merck
  • Resistance Risk Evaluated by Metaflumizone Selection and the Effects on Toxicities Over Other Insecticides in Spodoptera exigua (Lepidoptera: Noctuidae).
Resistance Risk Evaluated by Metaflumizone Selection and the Effects on Toxicities Over Other Insecticides in Spodoptera exigua (Lepidoptera: Noctuidae).
Journal of economic entomology (2019-06-21)
Xing-Xing Sun, Hong-Yang Li, Ying-Jie Jiang, Jun-Xi Zhang, Hui-Ling Gu, Bo Gao, Jing-Jing Ma, Fan Wang, Jia-Chun Zhou, Xiang-Rui Tian, Jianya Su, Kai Wang
ABSTRACT
Metaflumizone is a novel semicarbazone insecticide. It functions as a sodium channel blocker insecticide (SCBI) with excellent insecticidal activity on most economically important lepidopterous pests. This study assessed the resistance risk of Spodoptera exigua (Hübner) (Lepidoptera: Noctuidae) to metaflumizone in the laboratory and the effects of metaflumizone selection on toxicities to other insecticides. Spodoptera exigua collected from a field population at Huizhou in 2012 were successively challenged by metaflumizone to evaluate the risk of resistance evolution. Twelve generations of selection increased resistance to metaflumizone by 3.4-fold and threshold trait analysis revealed that the realized heritability (h2) of this resistance was 0.086. When h2 was equal to 0.086 and 90% of individuals were killed at each generation, LC50 to metaflumizone increased by 10-fold after 15 generations. The selection by metaflumizone did not increase the resistance to indoxacarb, chlorantraniliprole, spinosad, methomyl, or endosulfan, suggesting a lack of cross-resistance. However, metaflumizone challenge upheld the recession of resistance to emamectin benzoate, chlorfluazuron, and tebufenozide. The block of resistance drops by metaflumizone exposure implied a possible cross-resistance between metaflumizone and these three insecticides. These results contribute to integrated resistance management of S. exigua.
Product Description
Supelco
Metaflumizone, PESTANAL®, analytical standard
Supelco
Chlorantraniliprole, PESTANAL®, analytical standard