Merck
  • Isothiocyanate-Functionalized Mesoporous Silica Nanoparticles as Building Blocks for the Design of Nanovehicles with Optimized Drug Release Profile.
Isothiocyanate-Functionalized Mesoporous Silica Nanoparticles as Building Blocks for the Design of Nanovehicles with Optimized Drug Release Profile.
Nanomaterials (Basel, Switzerland) (2019-09-01)
Gabriel Martínez-Edo, Maria C Llinàs, Salvador Borrós, David Sánchez-García
ABSTRACT
A straightforward methodology for the synthesis of isothiocyanate-functionalized mesoporous silica nanoparticles (MSNs) by exposure of aminated MSNs to 1,1'-thiocarbonyldi-2(1H)-pyridone is reported. These nanoparticles are chemically stable, water tolerant, and readily react with primary amines without the formation of any by-product. This feature allows the easy modification of the surface of the nanoparticles for tuning their physical properties and the introduction of gatekeepers on the pore outlets. As a proof-of-concept, amino-isothiocyanate-functionalized MSNs have been used for the design of a nanocontainer able to release the drug Ataluren. The release profile of the drug can be easily fine-tuned with the careful choice of the capping amine.
Sigma-Aldrich
3-Bromopropionic acid, 97%
Sigma-Aldrich
(2-Aminoethyl)trimethylammonium chloride hydrochloride, 99%
Sigma-Aldrich
Tetraethyleneglycol monomethyl ether, technical grade