Merck
  • Evaluation of the Antifungal Activity of Mentha x piperita (Lamiaceae) of Pancalieri (Turin, Italy) Essential Oil and Its Synergistic Interaction with Azoles.
Evaluation of the Antifungal Activity of Mentha x piperita (Lamiaceae) of Pancalieri (Turin, Italy) Essential Oil and Its Synergistic Interaction with Azoles.
Molecules (Basel, Switzerland) (2019-09-01)
Vivian Tullio, Janira Roana, Daniela Scalas, Narcisa Mandras
ABSTRACT
The promising antimicrobial activity of essential oils (EOs) has led researchers to use them in combination with antimicrobial drugs in order to reduce drug toxicity, side effects, and resistance to single agents. Mentha x piperita, known worldwide as "Mentha of Pancalieri", is produced locally at Pancalieri (Turin, Italy). The EO from this Mentha species is considered as one of the best mint EOs in the world. In our research, we assessed the antifungal activity of "Mentha of Pancalieri" EO, either alone or in combination with azole drugs (fluconazole, itraconazole, ketoconazole) against a wide panel of yeast and dermatophyte clinical isolates. The EO was analyzed by GC-MS, and its antifungal properties were evaluated by minimum inhibitory concentration (MIC) and minimum fungicidal concentration (MFC) parameters, in accordance with the CLSI guidelines, with some modifications. The interaction of EO with azoles was evaluated through the chequerboard and isobologram methods. The results suggest that this EO exerts a fungicidal activity against yeasts and a fungistatic activity against dermatophytes. Interaction studies with azoles indicated mainly synergistic profiles between itraconazole and EO vs. Candida spp., Cryptococcus neoformans, and Trichophyton mentagrophytes. Thus, the "Mentha of Pancalieri" EO may act as a potential antifungal agent and could serve as a natural adjuvant for fungal infection treatment.
Sigma-Aldrich
Itraconazole, ≥98% (HPLC)
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
Menthone, mixture of isomers, ≥97.0% (GC)
Supelco
Menthone, analytical standard
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder