Merck
  • CENP-A reduction induces a p53-dependent cellular senescence response to protect cells from executing defective mitoses.
CENP-A reduction induces a p53-dependent cellular senescence response to protect cells from executing defective mitoses.
Molecular and cellular biology (2010-02-18)
Kayoko Maehara, Kohta Takahashi, Shigeaki Saitoh
ABSTRACT
Cellular senescence is an irreversible growth arrest and is presumed to be a natural barrier to tumor development. Like telomere shortening, certain defects in chromosome integrity can trigger senescence; however, the roles of centromere proteins in regulating commitment to the senescent state remains to be established. We examined chromatin structure in senescent human primary fibroblasts and found that CENP-A protein levels are diminished in senescent cells. Senescence-associated reduction of CENP-A is caused by transcriptional and posttranslational control. Surprisingly, forced reduction of CENP-A by short-hairpin RNA was found to cause premature senescence in human primary fibroblasts. This premature senescence is dependent on a tumor suppressor, p53, but not on p16(INK4a)-Rb; the depletion of CENP-A in p53-deficient cells results in aberrant mitosis with chromosome missegregation. We propose that p53-dependent senescence that arises from CENP-A reduction acts as a "self-defense mechanism" to prevent centromere-defective cells from undergoing mitotic proliferation that potentially leads to massive generation of aneuploid cells.
Sigma-Aldrich
Anti-p53 (Ab-6) (Pantropic) Mouse mAb (DO-1), liquid, clone DO-1, Calbiochem®
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-Heterochromatin Protein-1 β Antibody, clone 1MOD-1A9, ascites fluid, clone 1MOD-1A9, Chemicon®
Sigma-Aldrich
Anti-CENP-A Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Chk2 Antibody, clone 7, clone 7, Upstate®, from mouse
Sigma-Aldrich
Anti-Histone H2B Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-CENP-B Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-p16 (Ab-1) Mouse mAb (DCS-50.1/H4), liquid, clone DCS-50.1/H4, Calbiochem®