- Neuroprotective effects of non-steroidal anti-inflammatory drugs by direct scavenging of nitric oxide radicals.
Neuroprotective effects of non-steroidal anti-inflammatory drugs by direct scavenging of nitric oxide radicals.
Recently, it has been reported that inflammatory processes are associated with the pathophysiology of Alzheimer's disease and that treatment of non-steroidal anti-inflammatory drugs reduce the risk for Alzheimer's disease. In the present study, we examined nitric oxide radical quenching activity of non-steroidal anti-inflammatory drugs and steroidal drugs using our established direct in vitro nitric oxide radical detecting system by electron spin resonance spectrometry. The non-steroidal anti-inflammatory drugs, aspirin, mefenamic acid, indomethacin and ketoprofen directly and dose-dependently scavenged generated nitric oxide radicals. In experiments of nitric oxide radical donor, NOC18-induced neuronal damage, these four non-steroidal drugs significantly prevented the NOC18-induced reduction of cell viability and apoptotic nuclear changes in neuronal cells without affecting the induction of inducible nitric oxide synthase-like immunoreactivity. However, ibuprofen, naproxen or steroidal drugs, which had less or no scavenging effects in vitro, showed almost no protective effects against NOC18-induced cell toxicity. These results suggest that the protective effects of the former four non-steroidal anti-inflammatory drugs against apoptosis might be mainly due to their direct nitric oxide radical scavenging activities in neuronal cells. These direct NO. quenching activities represent novel effects of non-steroidal anti-inflammatory drugs. Our findings identified novel pharmacological mechanisms of these drugs to exert not only their anti-inflammatory, analgesic, antipyretic activities but also neuroprotective activities against neurodegeneration.