Merck
  • Morpho-taxonomic, genetic, and biochemical characterization of freshwater microalgae as potential biodiesel feedstock.
Morpho-taxonomic, genetic, and biochemical characterization of freshwater microalgae as potential biodiesel feedstock.
3 Biotech (2019-04-05)
Anisha Sehgal, Kongkana Goswami, Mintu Pal, Channakeshavaiah Chikkaputtaiah, Pankaj Chetia, Hari Prasanna Deka Boruah
ABSTRACT
In the present study, seven axenic fresh water microchlorophytes were isolated and identified as Tetradesmus dimorphus (NEIST BT-1), Chlorella sorokiniana (NEIST BT-2), Desmodesmus sp. (NEIST BT-10), Selenastrum sp. (NEIST BT-A6), Tetradesmus obliquus (NEIST BT-A1), Tetradesmus sp. (NEIST BT-A10), and Asterarcys sp. (NEIST BT-A15) based on morphological and molecular characterization. Their potential to be used as biodiesel feedstock was evaluated depending on their growth characteristics and lipid profiles. Among the seven isolates, NEIST BT-2 was found to be the most promising candidate owing to its high biomass yield (2.09 ± 0.037 g L-1) and lipid productivity (107.60 ± 10.175 mg L-1 day-1). The gas chromatography analysis confirmed the presence of significant amounts of palmitic acid, linoleic acid, linolenic acid, and oleic acid in the isolate which are some of the major constituents of any biodiesel. The predictive models showed that the biodiesel from this isolate has ideal fuel properties which comply with the ASTM D6751 and EN 14214 specifications. These findings demonstrate that NEIST BT-2 can be used as a prospective candidate for consideration of large-scale biodiesel production.