- Ln(OTf)3-catalysed highly regioselective alcoholysis of 2,3-epoxy alcohols.
Ln(OTf)3-catalysed highly regioselective alcoholysis of 2,3-epoxy alcohols.
Organic & biomolecular chemistry (2019-03-23)
Daichi Nakamura, Yusuke Sasano, Yoshiharu Iwabuchi
PMID30901005
ABSTRACT
Regioselectivity of Ln(OTf)3-catalysed alcoholysis of 2,3- and 3,4-epoxy alcohols was closely investigated to expand the scope of the transformations. The synthetic use was demonstrated by application to the construction of 4-propoxy-5-hydroxy-2,3-pentanedione (C4-propoxy-HPD), which is a potent synthetic mediator in AI-2 quorum sensing.