Merck
  • Sphingosine Kinase 1 Signaling Promotes Metastasis of Triple-Negative Breast Cancer.
Sphingosine Kinase 1 Signaling Promotes Metastasis of Triple-Negative Breast Cancer.
Cancer research (2019-06-27)
Sunil Acharya, Jun Yao, Ping Li, Chenyu Zhang, Frank J Lowery, Qingling Zhang, Hua Guo, Jingkun Qu, Fei Yang, Ignacio I Wistuba, Helen Piwnica-Worms, Aysegul A Sahin, Dihua Yu
ABSTRACT
Triple-negative breast cancer (TNBC) is the most aggressive breast cancer subtype. To identify TNBC therapeutic targets, we performed integrative bioinformatics analysis of multiple breast cancer patient-derived gene expression datasets and focused on kinases with FDA-approved or in-pipeline inhibitors. Sphingosine kinase 1 (SPHK1) was identified as a top candidate. SPHK1 overexpression or downregulation in human TNBC cell lines increased or decreased spontaneous metastasis to lungs in nude mice, respectively. SPHK1 promoted metastasis by transcriptionally upregulating the expression of the metastasis-promoting gene FSCN1 via NFκB activation. Activation of the SPHK1/NFκB/FSCN1 signaling pathway was associated with distance metastasis and poor clinical outcome in patients with TNBC. Targeting SPHK1 and NFκB using clinically applicable inhibitors (safingol and bortezomib, respectively) significantly inhibited aggressive mammary tumor growth and spontaneous lung metastasis in orthotopic syngeneic TNBC mouse models. These findings highlight SPHK1 and its downstream target, NFκB, as promising therapeutic targets in TNBC. SIGNIFICANCE: SPHK1 is overexpressed in TNBC and promotes metastasis, targeting SPHK1 or its downstream target NFκB with clinically available inhibitors could be effective for inhibiting TNBC metastasis.
Sigma-Aldrich
Anti-FSCN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%