Merck
A Code of Mono-phosphorylation Modulates the Function of RB.
Molecular cell (2019-02-04)
Ioannis Sanidas, Robert Morris, Katerina A Fella, Purva H Rumde, Myriam Boukhali, Eric C Tai, David T Ting, Michael S Lawrence, Wilhelm Haas, Nicholas J Dyson
ABSTRACT
Hyper-phosphorylation of RB controls its interaction with E2F and inhibits its tumor suppressor properties. However, during G1 active RB can be mono-phosphorylated on any one of 14 CDK phosphorylation sites. Here, we used quantitative proteomics to profile protein complexes formed by each mono-phosphorylated RB isoform (mP-RB) and identified the associated transcriptional outputs. The results show that the 14 sites of mono-phosphorylation co-ordinate RB's interactions and confer functional specificity. All 14 mP-RBs interact with E2F/DP proteins, but they provide different shades of E2F regulation. RB mono-phosphorylation at S811, for example, alters RB transcriptional activity by promoting its association with NuRD complexes. The greatest functional differences between mP-RBs are evident beyond the cell cycle machinery. RB mono-phosphorylation at S811 or T826 stimulates the expression of oxidative phosphorylation genes, increasing cellular oxygen consumption. These results indicate that RB activation signals are integrated in a phosphorylation code that determines the diversity of RB activity.
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder