MilliporeSigma
  • Preparation, Characterization, and Stability Evaluation of Taste-Masking Lacosamide Microparticles.
Preparation, Characterization, and Stability Evaluation of Taste-Masking Lacosamide Microparticles.
Materials (Basel, Switzerland) (2019-03-29)
Chang-Soo Han, Seungsu Kim, Dong-Won Oh, Jeong Yeol Yoon, Eun-Seok Park, Yun-Seok Rhee, Ju-Young Kim, Dae Hwan Shin, Dong-Wook Kim, Chun-Woong Park
ABSTRACT
Lacosamide (LCM) is a third-generation antiepileptic drug. Selective action of the drug on voltage-gated sodium channels reduces side effects. Oral administration of LCM shows good pharmacokinetic profile. However, the bitter taste of LCM is a barrier to the development of oral formulations. In this study, we aimed to prepare encapsulated LCM microparticles (MPs) for masking its bitter taste. Encapsulated LCM MPs were prepared with Eudragit® E100 (E100), a pH-dependent polymer, by spray drying. Three formulations comprising different ratios of LCM and E100 (3:1, 1:1, and 1:3) were prepared. Physicochemical tests showed that LCM was in an amorphous state in the prepared formulations, and they were not miscible. LCM-E100 (1:3) had a rough surface due to surface enrichment of LCM. Increased E100 ratio in LCM-E100 MPs resulted in better taste-making effectiveness: LCM-E100 (1:1) and LCM-E100 (1:3) showed good taste-masking effectiveness, while LCM-E100 (3:1) could not mask the bitter taste of LCM. Dissolution results of the prepared formulations showed good correlation with taste-masking effectiveness. Nevertheless, high E100 ratio reduced the stability of the prepared formulations. Especially the difference in initial dissolution profile observed for LCM-E100 (1:3) indicated rapid reduction in taste-masking effectiveness and surface recrystallization. Therefore, LCM-E100 formulation in the ratio of 1:1 was selected as the best formulation with good taste-masking effectiveness and stability.