- The Role of AMPARs in the Maturation and Integration of Caudal Ganglionic Eminence-Derived Interneurons into Developing Hippocampal Microcircuits.
The Role of AMPARs in the Maturation and Integration of Caudal Ganglionic Eminence-Derived Interneurons into Developing Hippocampal Microcircuits.
In the hippocampal CA1, caudal ganglionic eminence (CGE)-derived interneurons are recruited by activation of glutamatergic synapses comprising GluA2-containing calcium-impermeable AMPARs and exert inhibitory regulation of the local microcircuit. However, the role played by AMPARs in maturation of the developing circuit is unknown. We demonstrate that elimination of the GluA2 subunit (GluA2 KO) of AMPARs in CGE-derived interneurons, reduces spontaneous EPSC frequency coupled to a reduction in dendritic glutamatergic synapse density. Removal of GluA1&2&3 subunits (GluA1-3 KO) in CGE-derived interneurons, almost completely eliminated sEPSCs without further reducing synapse density, but increased dendritic branching. Moreover, in GluA1-3 KOs, the number of interneurons invading the hippocampus increased in the early postnatal period but converged with WT numbers later due to increased apoptosis. However, the CCK-containing subgroup increased in number, whereas the VIP-containing subgroup decreased. Both feedforward and feedback inhibitory input onto pyramidal neurons was decreased in GluA1-3 KO. These combined anatomical, synaptic and circuit alterations, were accompanied with a wide range of behavioural abnormalities in GluA1-3 KO mice compared to GluA2 KO and WT. Thus, AMPAR subunits differentially contribute to numerous aspects of the development and maturation of CGE-derived interneurons and hippocampal circuitry that are essential for normal behaviour.