Merck
  • Effects of GDNF-Transfected Marrow Stromal Cells on Rats with Intracerebral Hemorrhage.
Effects of GDNF-Transfected Marrow Stromal Cells on Rats with Intracerebral Hemorrhage.
Journal of stroke and cerebrovascular diseases : the official journal of National Stroke Association (2019-06-30)
Li Deng, Xiaoqing Gao, Guangbi Fan, Chaoxian Yang
ABSTRACT
The present study aimed to investigate the effects of Mesenchymal stem cells/glial cell line derived neurotrophic factor (MSCs/GDNF) transplantation on nerve reconstruction in rats with intracerebral hemorrhage. GDNF transduction to MSCs was using adenovirus vector pAdEasy-1-pAdTrack-CMV prepared. Intracerebral hemorrhage (ICH) was induced by injection of collagenase and heparin into the caudate putamen. At the third day after a collagenase-induced ICH, adult male SD rats were randomly divided into saline group, MSCs group and MSCs/GDNF group. Immunofluorescence and RT-PCR were performed to detect the differentiation of MSCs or MSCs with an adenovirus vector encoding GDNF gene in vivo and in vitro. After 6 hours of induction, both MSCs and MSCs/GDNF expressed neuro or glial specific markers and synaptic-associated proteins (SYN, GAP-43, PSD-95); additionally, they secreted bioactive compounds (BDNF, NGF-β). MSCs/GDNF transplantation, compared to MSCs and saline solution injection, significantly improved neurological functions after ICH. The grafted MSCs or MSCs/GDNF survived in the striatum after 2 weeks of transplantation and expressed the neural cell-specific biomarkers NSE, MAP2, and GFAP. These findings demonstrate that MSCs/GDNF transplantation contributes to improved neurological function in experimental ICH rats. The mechanisms are possibly due to neuronal replacement and enhanced neurotrophic factor secretion.