Merck
  • Proteomic profiling of HIV-1 infection of human CD4+ T cells identifies PSGL-1 as an HIV restriction factor.
Proteomic profiling of HIV-1 infection of human CD4+ T cells identifies PSGL-1 as an HIV restriction factor.
Nature microbiology (2019-03-06)
Ying Liu, Yajing Fu, Qian Wang, Mushan Li, Zheng Zhou, Deemah Dabbagh, Chunyan Fu, Hang Zhang, Shuo Li, Tengjiang Zhang, Jing Gong, Xiaohui Kong, Weiwei Zhai, Jiaming Su, Jianping Sun, Yonghong Zhang, Xiao-Fang Yu, Zhen Shao, Feng Zhou, Yuntao Wu, Xu Tan
ABSTRACT
Human immunodeficiency virus (HIV) actively modulates the protein stability of host cells to optimize viral replication. To systematically examine this modulation in HIV infection, we used isobaric tag-based mass spectrometry to quantify changes in the abundance of over 14,000 proteins during HIV-1 infection of human primary CD4+ T cells. We identified P-selectin glycoprotein ligand 1 (PSGL-1) as an HIV-1 restriction factor downregulated by HIV-1 Vpu, which binds to PSGL-1 and induces its ubiquitination and degradation through the ubiquitin ligase SCFβ-TrCP2. PSGL-1 is induced by interferon-γ in activated CD4+ T cells to inhibit HIV-1 reverse transcription and potently block viral infectivity by incorporating in progeny virions. This infectivity block is antagonized by Vpu via PSGL-1 degradation. We further show that PSGL-1 knockdown can significantly abolish the anti-HIV activity of interferon-γ in primary CD4+ T cells. Our study identifies an HIV restriction factor and a key mediator of interferon-γ's anti-HIV activity.