Merck
  • Hypertension-induced changes in the rat myocardium during the development of cardiac hypertrophy - a comparison between the left and the right ventricle.
Hypertension-induced changes in the rat myocardium during the development of cardiac hypertrophy - a comparison between the left and the right ventricle.
Acta histochemica (2018-10-20)
Alexandar Iliev, Georgi Kotov, Iva N Dimitrova, Boycho Landzhov
ABSTRACT
The hypertrophy of the cardiac muscle is one of the most significant maladaptive mechanisms activated in response to increased workload. It is associated with histological and ultrastructural alterations, changes in the quantitative parameters and the expression of different enzymes. While the structural and functional consequences of systemic hypertension on the left ventricle have been well evaluated, the right ventricle has received less attention. The aim of the present study was to analyse and compare the changes in the left and right ventricle during the development of cardiac hypertrophy initiated by systemic hypertension in different age groups of spontaneously hypertensive rats. Therefore, we studied the histology and ultrastructure of the cells of the myocardium, evaluated the immunohistochemical expression of the enzyme neuronal nitric oxide synthase and conducted a quantitative analysis of several morphometric parameters. We used three groups of spontaneously hypertensive rats. For the quantitative analysis we also used three age groups of age- and weight-matched control animals (normotensive Wistar rats). In both ventricles, we described cardiomyocytic hypertrophy, focal myocytolysis and increased collagen deposition in the interstitial space. Our observations on the ultrastructural level were associated with changes in the cardiomyocytic nuclei, the arrangement, maturity and organisation of the myofibrils, the localisation and ultrastructure of the mitochondria, the development and maturity of the intercalated discs, as well as changes in the components of the interstitium. The immunohistochemical expression of neuronal nitric oxide synthase in the left ventricle was stronger than that in the right ventricle across all age groups. The comparative quantitative analysis revealed that changes in the studied morphometric parameters in the two ventricles occurred disproportionately. In conclusion, the present study characterised the development of cardiac hypertrophy in response to systemic hypertension in both ventricles and demonstrated the involvement of the right ventricle.