Merck
  • Auto-regulation of Secretory Flux by Sensing and Responding to the Folded Cargo Protein Load in the Endoplasmic Reticulum.
Auto-regulation of Secretory Flux by Sensing and Responding to the Folded Cargo Protein Load in the Endoplasmic Reticulum.
Cell (2019-03-09)
Advait Subramanian, Anita Capalbo, Namrata Ravi Iyengar, Riccardo Rizzo, Antonella di Campli, Rosaria Di Martino, Matteo Lo Monte, Andrea R Beccari, Amol Yerudkar, Carmen Del Vecchio, Luigi Glielmo, Gabriele Turacchio, Marinella Pirozzi, Sang Geon Kim, Petra Henklein, Jorge Cancino, Seetharaman Parashuraman, Dario Diviani, Francesca Fanelli, Michele Sallese, Alberto Luini
ABSTRACT
Maintaining the optimal performance of cell processes and organelles is the task of auto-regulatory systems. Here we describe an auto-regulatory device that helps to maintain homeostasis of the endoplasmic reticulum (ER) by adjusting the secretory flux to the cargo load. The cargo-recruiting subunit of the coatomer protein II (COPII) coat, Sec24, doubles as a sensor of folded cargo and, upon cargo binding, acts as a guanine nucleotide exchange factor to activate the signaling protein Gα12 at the ER exit sites (ERESs). This step, in turn, activates a complex signaling network that activates and coordinates the ER export machinery and attenuates proteins synthesis, thus preventing large fluctuations of folded and potentially active cargo that could be harmful to the cell or the organism. We call this mechanism AREX (autoregulation of ER export) and expect that its identification will aid our understanding of human physiology and diseases that develop from secretory dysfunction.
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
PKA Inhibitor 14-22 Amide, Cell-Permeable, Myristoylated, PKA Inhibitor 14-22 Amide is myristoylated at the N-terminus that enhances its cell-permeability. The non-myristoylated version is shown to be a specific inhibitor of PKA (Ki = 36 nM).
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Millipore
ANTI-FLAG® High Sensitivity, M2 coated 96-well plates, 96-well, clear, polystyrene, flat bottom plate
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-PDE3B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
Forskolin, Coleus forskohlii
Sigma-Aldrich
Anti-VSV Glycoprotein antibody, Mouse monoclonal, clone P5D4, purified from hybridoma cell culture