Merck
  • Acute insulin signaling in pancreatic beta-cells is mediated by multiple Raf-1 dependent pathways.
Acute insulin signaling in pancreatic beta-cells is mediated by multiple Raf-1 dependent pathways.
Endocrinology (2010-01-09)
Emilyn U Alejandro, Tatyana B Kalynyak, Farnaz Taghizadeh, Kamila S Gwiazda, Erin K Rawstron, Karen J Jacob, James D Johnson
ABSTRACT
Insulin enhances the proliferation and survival of pancreatic beta-cells, but its mechanisms remain unclear. We hypothesized that Raf-1, a kinase upstream of both ERK and Bad, might be a critical target of insulin in beta-cells. To test this hypothesis, we treated human and mouse islets as well as MIN6 beta-cells with multiple insulin concentrations and examined putative downstream targets using immunoblotting, immunoprecipitation, quantitative fluorescent imaging, and cell death assays. Low doses of insulin rapidly activated Raf-1 by dephosphorylating serine 259 and phosphorylating serine 338 in human islets, mouse islets, and MIN6 cells. The phosphorylation of ERK by insulin was eliminated by exposure to a Raf inhibitor (GW5074) or transfection with a dominant-negative Raf-1 mutant. Insulin also enhanced the interaction between mitochondrial Raf-1 and Bcl-2 agonist of cell death (Bad), promoting Bad inactivation via its phosphorylation on serine 112. Insulin-stimulated ERK phosphorylation was abrogated by calcium chelation, calcineurin and calmodulin-dependent protein kinase II inhibitors, and Ned-19, a nicotinic acid adenine dinucleotide phosphate receptor (NAADPR) antagonist. Blocking Raf-1 and Ca(2+) signaling resulted in nonadditive beta-cell death. Autocrine insulin signaling partly accounted for the effects of glucose on ERK phosphorylation. Our results demonstrate that Raf-1 is a critical target of insulin in primary beta-cells. Activation of Raf-1 leads to both an ERK-dependent pathway that involves nicotinic acid adenine dinucleotide phosphate-sensitive Ca(2+) stores and Ca(2+)-dependent phosphorylation events, and an ERK-independent pathway that involves Bad inactivation at the mitochondria. Together our findings identify a novel insulin signaling pathway in beta-cells and shed light on insulin's antiapoptotic and mitogenic mechanisms.
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)
Sigma-Aldrich
Mitochondria Isolation Kit, sufficient for 10-20 g (animal tissue), sufficient for 50 assays (2 mL), isolation of enriched mitochondrial fraction from animal tissues