Merck
  • Antimycobacterial spectrum of sparfloxacin and its activities alone and in association with other drugs against Mycobacterium avium complex growing extracellularly and intracellularly in murine and human macrophages.
Antimycobacterial spectrum of sparfloxacin and its activities alone and in association with other drugs against Mycobacterium avium complex growing extracellularly and intracellularly in murine and human macrophages.
Antimicrobial agents and chemotherapy (1991-12-01)
N Rastogi, V Labrousse, K S Goh, J P De Sousa
ABSTRACT
The MICs and MBCs of the new difluorinated quinolone drug sparfloxacin against type strains belonging to 21 species of mycobacteria were screened. The MICs and MBCs were within the range of 0.1 to 2.0 and 0.1 to 4.0 micrograms/ml, respectively (with an MBC/MIC ratio of 1 to 2), and against 18 of the 21 species tested, the drug showed significant bactericidal activity (at least 99% killing or more of the initial inoculum added) at concentrations well within the reported peak concentrations in serum (Cmax) in humans. MICs of sparfloxacin for 7 of 10 Mycobacterium avium complex strains were below the Cmax, with MBC/MIC ratios within the range of 2 to 4. Enhancement of its activity by ethambutol, rifampin, amikacin, and clarithromycin (which were used at sublethal concentrations) assessed by using BACTEC radiometry revealed that its activity was further enhanced in 2 of 10 strains by rifampin and in 7 of 10 strains by ethambutol. The bactericidal effects of various drugs used alone as well as two-drug combinations used at Cmax levels were also screened against four strains of M. avium complex growing intracellularly in two different macrophage systems, namely, mouse bone marrow-derived macrophages and peripheral blood monocyte-derived human macrophages. Our results showed a satisfactory correlation between the extracellular and intracellular drug activity data.
Sigma-Aldrich
Sparfloxacin, ≥98.0% (HPLC)