Merck
  • MHC class II compartment, endocytosis and phagocytic activity of macrophages and putative dendritic cells isolated from normal tissues rich in synovium.
MHC class II compartment, endocytosis and phagocytic activity of macrophages and putative dendritic cells isolated from normal tissues rich in synovium.
International immunology (2005-07-20)
Mahin Moghaddami, Graham Mayrhofer, Leslie G Cleland
ABSTRACT
The endocytic and phagocytic activities of a population of MHC IIhi CD11c+ dendritic cell (DC)-like cells in synovium-rich tissues (SRTs) of normal rat paws were compared with CD163+ cells (putative macrophages) from the same tissues and pseudo-afferent lymph DCs, peritoneal macrophages and blood monocytes. Fifty percent of CD11c+ cells and 75% of CD163+ cells isolated from SRT internalized fluorescein-conjugated dextran (FITC-DX). Of these endocytic cells, half of those expressing CD11c, but only 30% of those expressing CD163, were surface MHC class II+ (sMHC II+). CD11c+ cells were more endocytic than monocytes or pseudo-afferent lymph DC, but some CD163+ cells (type A synoviocytes) were found to be highly endocytic. CD163+ cells from SRT were more phagocytic (25%) than the general MHC class II+ population (16%). Of phagocytic cells, 40% of CD163+ cells were sMHC II(variable) and they constituted 60% of all MHC class II+ phagocytic cells. Only 18% of phagocytic MHC II+ cells expressed CD11c and the most of these were MHC IIhi. In comparison, 60% of CD163+ peritoneal macrophages were phagocytic, while blood monocytes were poorly phagocytic. Intracellular MHC class II-rich compartments (MIIC) were prominent in sMHC IIhi cells in SRT but rare in CD163+ cells. Most MHC IIhi CD11c+ cells did not have a detectable MIIC.
Sigma-Aldrich
Latex beads, carboxylate-modified polystyrene, fluorescent red, aqueous suspension, 0.5 μm mean particle size